• USA Home
  • 77445 - 2-Aminoacetophenone hydrochloride

EMAIL THIS PAGE TO A FRIEND
77445 Sigma-Aldrich

2-Aminoacetophenone hydrochloride

purum, ≥98.0% (AT)

Synonym: ω-Aminoacetophenone hydrochloride, Phenacylamine hydrochloride

  • CAS Number 5468-37-1

  • Linear Formula C6H5COCH2NH2 · HCl

  • Molecular Weight 171.62

  •  Beilstein Registry Number 3563173

  •  EC Number 226-787-2

  •  MDL number MFCD00012873

  •  PubChem Substance ID 24887193

Purchase

Properties

Related Categories Building Blocks, C7 to C8, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   CVXGFPPAIUELDV-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)
mp   194 °C (dec.)(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?