EMAIL THIS PAGE TO A FRIEND
77624 Sigma-Aldrich

Dexpanthenol

tested according to Ph.Eur.

Synonym: (R)-(+)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide, (R)-2,4-Dihydroxy-3,3-dimethylbutyric 3-hydroxypropylamide, D-Panthenol, D-Pantothenyl alcohol, Dexpanthenol, Dexpanthenolum, Provitamin B

  • CAS Number 81-13-0

  • Empirical Formula (Hill Notation) C9H19NO4

  • Molecular Weight 205.25

  •  Beilstein Registry Number 1724947

  •  EC Number 201-327-3

  •  MDL number MFCD00065006

  •  PubChem Substance ID 329767823

Purchase

Properties

Related Categories Analytical/Chromatography, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Core Bioreagents,
grade   Ph Eur (tested according to)
InChI Key   SNPLKNRPJHDVJA-ZETCQYMHSA-N
form   viscous liquid (clear)
pharmacopeia   testing conforms to Pharmacopeia
density   1.20 g/mL at 20 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?