• USA Home
  • 77710 - 3-Phenoxybenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
77710 Sigma-Aldrich

3-Phenoxybenzyl alcohol

purum, ≥97.5% (GC)

  • CAS Number 13826-35-2

  • Linear Formula C6H5OC6H4CH2OH

  • Molecular Weight 200.23

  •  Beilstein Registry Number 475312

  •  EC Number 237-525-1

  •  MDL number MFCD00004636

Purchase

Properties

Related Categories Alcohols, Building Blocks, C11 to C30+, Chemical Synthesis, Organic Building Blocks,
vapor pressure   0.1 mmHg ( 37.7 °C)
InChI Key   KGANAERDZBAECK-UHFFFAOYSA-N
grade   purum
assay   ≥97.5% (GC)
refractive index   n20/D 1.591(lit.)
  n20/D 1.591
bp   135-140 °C/0.1 mmHg(lit.)
density   1.149 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
DP0775400
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

190284 3-Phenoxybenzyl alcohol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?