• USA Home
  • 77800 - Phenylacetaldehyde solution

EMAIL THIS PAGE TO A FRIEND
77800 Sigma-Aldrich

Phenylacetaldehyde solution

technical, ~50% in diethyl phthalate

  • CAS Number 122-78-1

  • Linear Formula C6H5CH2CHO

  • Molecular Weight 120.15

  •  Beilstein Registry Number 385791

  •  MDL number MFCD00006993

  •  PubChem Substance ID 329767941

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C8, Carbonyl Compounds, Chemical Synthesis,
InChI Key   DTUQWGWMVIHBKE-UHFFFAOYSA-N
grade   technical
concentration   ~50% in diethyl phthalate
impurities   ≤2% free acid
refractive index   n20/D 1.521
density   1.079 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
2
Flash Point(F) 
188.6 °F
Flash Point(C) 
87 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?