• USA Home
  • 77845 - (S)-(+)-1-Phenyl-1,2-ethanediol

EMAIL THIS PAGE TO A FRIEND
77845 Sigma-Aldrich

(S)-(+)-1-Phenyl-1,2-ethanediol

purum, ≥98.0% (sum of enantiomers, HPLC)

Synonym: (+)-Styrene glycol, (S)-(+)-Phenylethylene glycol

  • CAS Number 25779-13-9

  • Linear Formula HOCH2CH(C6H5)OH

  • Molecular Weight 138.16

  •  Beilstein Registry Number 3196197

  •  MDL number MFCD00066256

  •  PubChem Substance ID 329767968

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Polyols More...
InChI Key   PWMWNFMRSKOCEY-MRVPVSSYSA-N
grade   purum
assay   ≥98.0% (sum of enantiomers, HPLC)
optical activity   [α]/D +69±4°, c = 1 in chloroform
mp   64-67 °C(lit.)
  64-67 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
320 °F
Flash Point(C) 
160 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?