• USA Home
  • 77847 - (R)-(−)-1-Phenyl-1,2-ethanediol

EMAIL THIS PAGE TO A FRIEND
77847 Sigma-Aldrich

(R)-(−)-1-Phenyl-1,2-ethanediol

purum, ≥98.0% (sum of enantiomers, HPLC)

Synonym: (−)-Styrene glycol, (R)-(−)-Phenylethylene glycol

  • CAS Number 16355-00-3

  • Linear Formula HOCH2CH(C6H5)OH

  • Molecular Weight 138.16

  •  Beilstein Registry Number 2355547

  •  MDL number MFCD00064262

  •  PubChem Substance ID 329767970

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Polyols More...
InChI Key   PWMWNFMRSKOCEY-QMMMGPOBSA-N
grade   purum
assay   ≥98.0% (sum of enantiomers, HPLC)
optical activity   [α]/D −69±4°, c = 1 in chloroform
bp   272-274 °C(lit.)
mp   64-67 °C(lit.)

Description

Other Notes

Chiral building block used in various syntheses

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

302163 (R)-(−)-1-Phenyl-1,2-ethanediol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?