• USA Home
  • 77870 - (S)-(−)-α-Methylbenzylamine

EMAIL THIS PAGE TO A FRIEND
77870 Sigma-Aldrich

(S)-(−)-α-Methylbenzylamine

purum, ≥98.0% (sum of enantiomers, GC)

Synonym: (S)-(−)-1-Phenylethylamine, (S)-(-)-alpha-Methylbenzylamine

Purchase

Properties

Related Categories Amines, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
vapor pressure   0.5 mmHg ( 20 °C)
InChI Key   RQEUFEKYXDPUSK-ZETCQYMHSA-N
grade   purum
assay   ≥98.0% (sum of enantiomers, GC)
optical activity   [α]20/D −30±2°, c = 10% in ethanol
optical purity   enantiomeric ratio: ≥97:3 (GC)
refractive index   n20/D 1.526(lit.)
  n20/D 1.528
bp   187 °C(lit.)
density   0.94 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2922 6.1(8) / PGII
WGK Germany 
1
Flash Point(F) 
158 °F
Flash Point(C) 
70 °C
Protocols & Articles

Articles

HPLC Analysis of α-Methylbenzylamine Enantiomers (DANSYL Derivatives) on Astec® CYCLOBOND® I 2000 DMP

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of α-Methylbenzylamine Enantiomers (DANSYL Derivatives) on Astec® CYCLOBOND® I 2000 DMP
Keywords: Chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?