• USA Home
  • 77920 - (2-Bromoethyl)benzene

EMAIL THIS PAGE TO A FRIEND
77920 Sigma-Aldrich

(2-Bromoethyl)benzene

purum, ≥97.0% (GC)

Synonym: 2-Phenylethyl bromide

  • CAS Number 103-63-9

  • Linear Formula C6H5CH2CH2Br

  • Molecular Weight 185.06

  •  Beilstein Registry Number 507487

  •  EC Number 203-130-8

  •  MDL number MFCD00000240

Purchase

Properties

Related Categories Aryl, Building Blocks, C8, Chemical Synthesis, Halogenated Hydrocarbons,
InChI Key   WMPPDTMATNBGJN-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.556(lit.)
  n20/D 1.556
bp   220-221 °C(lit.)
density   1.355 g/mL at 25 °C(lit.)

Description

Caution

may discolor to yellow on storage

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993 / PGIII
WGK Germany 
3
RTECS 
CY9032000
Flash Point(F) 
204.8 °F
Flash Point(C) 
96 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

B65780 (2-Bromoethyl)benzene, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?