EMAIL THIS PAGE TO A FRIEND
78190 Sigma-Aldrich

Phenylpyruvic acid

purum, ≥98.0% (T)

Synonym: 2-Oxo-3-phenylpropanoic acid, 2-Oxo-3-phenylpropionic acid

  • CAS Number 156-06-9

  • Linear Formula C6H5CH2COCOOH

  • Molecular Weight 164.16

  •  Beilstein Registry Number 2207312

  •  EC Number 205-847-1

  •  MDL number MFCD00002589

Purchase

Properties

Related Categories Building Blocks, C9, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   BTNMPGBKDVTSJY-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
mp   150-154 °C(lit.)
  150-154 °C
storage temp.   −20°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

286958 Phenylpyruvic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?