• USA Home
  • 78195 - Calcium phenylpyruvate

EMAIL THIS PAGE TO A FRIEND
78195 Sigma-Aldrich

Calcium phenylpyruvate

purum, ≥98.0% (dry substance, NT)

Synonym: Phenylpyruvic acid calcium salt

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Carboxylic Acid Salts, Chemical Synthesis, Organic Building Blocks More...
InChI Key   BKMMOJVSLBZNTK-UHFFFAOYSA-L
grade   purum
assay   ≥98.0% (dry substance, NT)
impurities   ≤6% water

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?