• USA Home
  • 78243 - (±)-3-Phenylbutyric acid

EMAIL THIS PAGE TO A FRIEND
78243 Sigma-Aldrich

(±)-3-Phenylbutyric acid

purum, ≥97.0% (GC)

Synonym: (±)-β-Methylhydrocinnamic acid

  • CAS Number 4593-90-2

  • Linear Formula CH3CH(C6H5)CH2COOH

  • Molecular Weight 164.20

  •  Beilstein Registry Number 2044322

  •  EC Number 224-987-4

  •  MDL number MFCD00002725

  •  PubChem Substance ID 24887279

Purchase

Properties

Related Categories Building Blocks, C10, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   ZZEWMYILWXCRHZ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
bp   170-172 °C/20 mmHg(lit.)
mp   35-38 °C(lit.)
  35-38 °C
density   1.515 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

116807 3-Phenylbutyric acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?