• USA Home
  • 78323 - (1S,2R)-trans-2-Phenyl-1-cyclohexanol

EMAIL THIS PAGE TO A FRIEND
78323 Sigma-Aldrich

(1S,2R)-trans-2-Phenyl-1-cyclohexanol

puriss., ≥99.0% (sum of enantiomers, GC)

  • CAS Number 34281-92-0

  • Empirical Formula (Hill Notation) C12H16O

  • Molecular Weight 176.25

  •  Beilstein Registry Number 4669439

  •  MDL number MFCD00066432

  •  PubChem Substance ID 24887287

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   AAIBYZBZXNWTPP-NEPJUHHUSA-N
grade   puriss.
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]20/D +59±2°, c = 10% in methanol
optical purity   enantiomeric ratio: ≥99:1 (GC)
mp   63-64 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?