EMAIL THIS PAGE TO A FRIEND
78366 Sigma-Aldrich

1-Phenyldecane

analytical standard

Synonym: Decylbenzene

  • CAS Number 104-72-3

  • Linear Formula C6H5(CH2)9CH3

  • Molecular Weight 218.38

  •  Beilstein Registry Number 1907745

  •  EC Number 203-230-1

  •  MDL number MFCD00008955

  •  PubChem Substance ID 24887292

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Aromatics,
grade   analytical standard
InChI Key   UZILCZKGXMQEQR-UHFFFAOYSA-N
assay   ≥99.5% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.482(lit.)
  n20/D 1.483
bp   293 °C(lit.)
mp   −14 °C(lit.)
density   0.856 g/mL at 25 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
UN 3082 9/PG 3
WGK Germany 
1
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?