EMAIL THIS PAGE TO A FRIEND
78410 Sigma-Aldrich

o-Phenylenediamine

suitable for fluorescence, ≥99.0% (NT)

Synonym: 1,2-Diaminobenzene, 1,2-Phenylenediamine, OPD

  • CAS Number 95-54-5

  • Empirical Formula (Hill Notation) C6H8N2

  • Molecular Weight 108.14

  •  Beilstein Registry Number 606074

  •  EC Number 202-430-6

  •  MDL number MFCD00007721

  •  PubChem Substance ID 329768144

Purchase

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 1673 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
276.8 °F
Flash Point(C) 
136 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P9029 o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?