EMAIL THIS PAGE TO A FRIEND
78422 Sigma-Aldrich

m-Phenylenediamine

technical, ≥98.0% (NT)

Synonym: 1,3-Benzenediamine, 1,3-Diaminobenzene, 1,3-Phenylenediamine, MPDA

  • CAS Number 108-45-2

  • Linear Formula C6H4-1,3-(NH2)2

  • Molecular Weight 108.14

  •  Beilstein Registry Number 471357

  •  EC Number 203-584-7

  •  MDL number MFCD00007799

  •  PubChem Substance ID 329768147

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Polyamines More...
vapor density   3.7 (vs air)
vapor pressure   0.62 mmHg ( 100 °C)
InChI Key   WZCQRUWWHSTZEM-UHFFFAOYSA-N
grade   technical
assay   ≥98.0% (NT)
autoignition temp.   1040 °F
bp   282-284 °C
mp   61-65 °C
  64-66 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 1673 6.1 / PGIII
WGK Germany 
3
RTECS 
SS7700000
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P23954 m-Phenylenediamine, flakes, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?