EMAIL THIS PAGE TO A FRIEND
78430 Sigma-Aldrich

p-Phenylenediamine

technical, ≥97.0% (GC/NT)

Synonym: 1,4-Benzenediamine, 1,4-Diaminobenzene, 1,4-Phenylenediamine

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Polyamines More...
vapor density   3.7 (vs air)
vapor pressure   1.08 mmHg ( 100 °C)
description   anti-fade reagent
InChI Key   CBCKQZAAMUWICA-UHFFFAOYSA-N
grade   technical
assay   ≥97.0% (GC/NT)
bp   267 °C(lit.)
mp   138-143 °C(lit.)
  138-143 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Target organs 
Heart, Kidney, Musculo-skeletal system
RIDADR 
UN 1673 6.1 / PGIII
WGK Germany 
3
RTECS 
SS8050000
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P6001 p-Phenylenediamine

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?