CAS Number 87-32-1
Empirical Formula (Hill Notation) C13H14N2O3
Molecular Weight 246.26
Beilstein Registry Number 89478
EC Number 201-739-3
MDL number MFCD00005644
| Quality Level | GMP |
| assay | 99.0-101.0% (w/w) |
| optical activity | [α]/D -0.1 to +0.1, c = 1 g/10 mL in 1 M NaOH |
| impurities | ≤10 mg/kg heavy metals |
| ign. residue | ≤0.1% |
| loss | ≤0.5% loss on drying |
| mp | 204-206 °C (dec.)(lit.) |
| solubility | 1 M NaOH: soluble0.1 g/10 mL, clear, colorless to almost colorless |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?