EMAIL THIS PAGE TO A FRIEND
78560 Sigma-Aldrich

N-Phenylglycine

purum, ≥97.0% (T)

Synonym: (Phenylamino)acetic acid, Anilinoacetic acid

  • CAS Number 103-01-5

  • Linear Formula C6H5NHCH2COOH

  • Molecular Weight 151.16

  •  Beilstein Registry Number 509838

  •  EC Number 203-070-2

  •  MDL number MFCD00014009

  •  PubChem Substance ID 329768160

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylglycine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   NPKSPKHJBVJUKB-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (T)
mp   121-123 °C(lit.)
  121-125 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

330469 N-Phenylglycine, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?