• USA Home
  • 78565 - L-(+)-α-Phenylglycine

EMAIL THIS PAGE TO A FRIEND
78565 Sigma-Aldrich

L-(+)-α-Phenylglycine

puriss., ≥99.0% (NT)

Synonym: (S)-(+)-α-Aminophenylacetic acid

  • CAS Number 2935-35-5

  • Empirical Formula (Hill Notation) C8H9NO2

  • Molecular Weight 151.16

  •  Beilstein Registry Number 2208675

  •  EC Number 220-909-8

  •  MDL number MFCD00064403

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylglycine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   ZGUNAGUHMKGQNY-ZETCQYMHSA-N
grade   puriss.
assay   ≥99.0% (NT)
optical activity   [α]20/D +156±2°, c = 1% in 1 M HCl

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

237647 L−(+)-α-Phenylglycine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?