• USA Home
  • 78570 - D-(−)-α-Phenylglycine

EMAIL THIS PAGE TO A FRIEND
78570 Sigma-Aldrich

D-(−)-α-Phenylglycine

puriss., ≥99.0% (NT)

Synonym: (R)-(−)-α-Aminophenylacetic acid

  • CAS Number 875-74-1

  • Empirical Formula (Hill Notation) C8H9NO2

  • Molecular Weight 151.16

  •  Beilstein Registry Number 2208676

  •  EC Number 212-876-3

  •  MDL number MFCD00008061

  •  PubChem Substance ID 329768163

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylglycine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   ZGUNAGUHMKGQNY-SSDOTTSWSA-N
grade   puriss.
assay   ≥99.0% (NT)
optical activity   [α]20/D −156±2°, c = 1% in 1 M HCl
ign. residue   ≤0.05%
mp   ≥300 °C (dec.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
302 °F
Flash Point(C) 
150 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

SPE tube or plate

Product #

Description

Add to Cart

P25485 D−(−)-α-Phenylglycine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?