EMAIL THIS PAGE TO A FRIEND
78580 Sigma-Aldrich

DL-α-Phenylglycine

purum, ≥98.0% (NT)

Synonym: (±)-α-Aminophenylacetic acid

  • CAS Number 2835-06-5

  • Linear Formula C6H5CH(NH2)COOH

  • Molecular Weight 151.16

  •  Beilstein Registry Number 3197862

  •  EC Number 220-608-1

  •  MDL number MFCD00064402

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylglycine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   ZGUNAGUHMKGQNY-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (NT)
mp   290 °C (subl.)(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

P25507 2-Phenylglycine, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?