EMAIL THIS PAGE TO A FRIEND
78670 Sigma-Aldrich

Phenylhydrazine

purum p.a., ≥95.0% (GC)

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Building Blocks, Chemical Synthesis, Hydrazines,
vapor density   4.3 (vs air)
vapor pressure   <0.1 mmHg ( 20 °C)
InChI Key   HKOOXMFOFWEVGF-UHFFFAOYSA-N
grade   purum p.a.
assay   ≥95.0% (GC)
autoignition temp.   345 °F
refractive index   n20/D 1.607(lit.)
bp   238-241 °C(lit.)
mp   18-21 °C(lit.)
density   1.098 g/mL at 25 °C(lit.)

Description

Application

for spectrophotometric determination of Al, Cr, Mo, Ti, Zr

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 2572 6.1 / PGII
WGK Germany 
3
RTECS 
MV8925000
Flash Point(F) 
192.2 °F
Flash Point(C) 
89 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P26252 Phenylhydrazine, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?