EMAIL THIS PAGE TO A FRIEND
78672 Sigma-Aldrich

Phenylhydrazine

purum, ≥95.0% (GC)

  • CAS Number 100-63-0

  • Linear Formula C6H5NHNH2

  • Molecular Weight 108.14

  •  Beilstein Registry Number 606080

  •  EC Number 202-873-5

  •  MDL number MFCD00007573

  •  PubChem Substance ID 24887154

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Hydrazines, Nitrogen Compounds, Organic Building Blocks More...
vapor density   4.3 (vs air)
vapor pressure   <0.1 mmHg ( 20 °C)
InChI Key   HKOOXMFOFWEVGF-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
autoignition temp.   345 °F
refractive index   n20/D 1.607(lit.)
bp   238-241 °C(lit.)
mp   18-21 °C(lit.)
density   1.098 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 2572 6.1 / PGII
WGK Germany 
3
RTECS 
MV8925000
Flash Point(F) 
192.2 °F
Flash Point(C) 
89 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?