• USA Home
  • 78681 - Pilocarpine nitrate salt

EMAIL THIS PAGE TO A FRIEND
78681 Sigma-Aldrich

Pilocarpine nitrate salt

tested according to Ph.Eur.

Synonym: Pilocarpini nitras

  • CAS Number 148-72-1

  • Empirical Formula (Hill Notation) C11H16N2O2 · HNO3

  • Molecular Weight 271.27

  •  Beilstein Registry Number 4171406

  •  EC Number 205-723-7

  •  MDL number MFCD00078497

  •  PubChem Substance ID 329768173

Purchase

Properties

Related Categories Analytical/Chromatography, Pharmacopoeia, Pharmacopoeia A-Z More...
InChI Key   PRZXEPJJHQYOGF-GNAZCLTHSA-N
pharmacopeia   testing conforms to Pharmacopeia

Description

Biochem/physiol Actions

Nonselective muscarinic acetylcholine receptor agonist; used to produce an experimental model of epilepsy.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?