EMAIL THIS PAGE TO A FRIEND
78795 Sigma-Aldrich

N-Phenylmaleimide

≥98.0% (HPLC)

  • CAS Number 941-69-5

  • Empirical Formula (Hill Notation) C10H7NO2

  • Molecular Weight 173.17

  •  Beilstein Registry Number 125098

  •  EC Number 213-382-0

  •  MDL number MFCD00005502

  •  PubChem Substance ID 24887368

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Chemical Synthesis, Cyclic Imides, Organic Building Blocks More...
InChI Key   HIDBROSJWZYGSZ-UHFFFAOYSA-N
assay   ≥98.0% (HPLC)
bp   162-163 °C/12 mmHg(lit.)
mp   85-87 °C(lit.)
  89-90 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2811 - class 6.1 - PG 3 - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
ON5950000
Flash Point(F) 
305.6 °F
Flash Point(C) 
152 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

P27100 N-Phenylmaleimide, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?