• USA Home
  • 78923 - (S)-(−)-1-Phenyl-1-propanol

EMAIL THIS PAGE TO A FRIEND
78923 Sigma-Aldrich

(S)-(−)-1-Phenyl-1-propanol

puriss., ≥99.0% (sum of enantiomers, GC)

Synonym: (S)-(−)-α-Ethylbenzyl alcohol

  • CAS Number 613-87-6

  • Linear Formula C2H5CH(C6H5)OH

  • Molecular Weight 136.19

  •  Beilstein Registry Number 2041556

  •  MDL number MFCD00066207

Purchase

Properties

Related Categories Alcohols, Analytical Reagents, Analytical/Chromatography, Asymmetric Synthesis, Chemical Synthesis,
InChI Key   DYUQAZSOFZSPHD-VIFPVBQESA-N
grade   puriss.
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]20/D −47±1°, c = 2.2% in hexane
optical purity   enantiomeric ratio: ≥99:1 (GC)
refractive index   n20/D 1.520(lit.)
  n20/D 1.520
bp   94-95 °C/10 mmHg(lit.)
density   0.992 g/mL at 25 °C(lit.)

Description

Application

Chiral building block

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
UN 2810 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
194 °F
Flash Point(C) 
90 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?