• USA Home
  • 79120 - Phenylmercury acetate

EMAIL THIS PAGE TO A FRIEND
79120 Sigma-Aldrich

Phenylmercury acetate

purum, ≥98.0% (Hg)

Synonym: Mercury phenyl acetate

  • CAS Number 62-38-4

  • Linear Formula C6H5HgOCOCH3

  • Molecular Weight 336.74

  •  Beilstein Registry Number 3662930

  •  EC Number 200-532-5

  •  MDL number MFCD00008691

  •  PubChem Substance ID 329768260

Purchase

Properties

Related Categories Chemical Synthesis, Organomercury, Organometallic Reagents More...
InChI Key   XEBWQGVWTUSTLN-UHFFFAOYSA-M
grade   purum
assay   ≥98.0% (Hg)
mp   148-151 °C(lit.)
  148-151 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN 1674 6.1 / PGII
WGK Germany 
3
RTECS 
OV6475000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P27127 Phenylmercuric acetate, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?