• USA Home
  • 79132 - Phenylmercury chloride

EMAIL THIS PAGE TO A FRIEND
79132 Sigma-Aldrich

Phenylmercury chloride

purum, ≥95.0% (Hg)

Purchase

Properties

Related Categories Chemical Synthesis, Organomercury, Organometallic Reagents More...
InChI Key   AWGTVRDHKJQFAX-UHFFFAOYSA-M
grade   purum
assay   ≥95.0% (Hg)
mp   248-250 °C (dec.)(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?