• USA Home
  • 79150 - 4-Phenylsemicarbazide

EMAIL THIS PAGE TO A FRIEND
79150 Sigma-Aldrich

4-Phenylsemicarbazide

puriss., for the detection of aldehydes and ketones, ≥98.5% (N)

  • CAS Number 537-47-3

  • Linear Formula C6H5NHCONHNH2

  • Molecular Weight 151.17

  •  Beilstein Registry Number 511107

  •  EC Number 208-669-2

  •  MDL number MFCD00007590

  •  PubChem Substance ID 329768275

Purchase

Properties

Related Categories Aldehydes, Analytical Reagents, Analytical/Chromatography, Building Blocks, Carbonyl Compounds,
InChI Key   MOCKWYUCPREFCZ-UHFFFAOYSA-N
grade   puriss.
assay   ≥98.5% (N)
quality   for the detection of aldehydes and ketones
mp   122-125 °C(lit.)
  123-127 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?