• USA Home
  • 79200 - 4-Phenylthiosemicarbazide

EMAIL THIS PAGE TO A FRIEND
79200 Sigma-Aldrich

4-Phenylthiosemicarbazide

puriss. p.a., for spectrophotometric det. of Se, Te, ≥99.0% (NT)

  • CAS Number 5351-69-9

  • Linear Formula C6H5NHCSNHNH2

  • Molecular Weight 167.23

  •  Beilstein Registry Number 608285

  •  EC Number 226-329-1

  •  MDL number MFCD00007615

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Building Blocks, Chemical Synthesis, Organic Building Blocks,
InChI Key   KKIGUVBJOHCXSP-UHFFFAOYSA-N
grade   puriss. p.a.
assay   ≥99.0% (NT)
quality   for spectrophotometric det. of Se, Te
ign. residue   ≤0.05%
mp   138-140 °C(lit.)
  138-140 °C
solubility   methanol: soluble50 mg/mL, clear

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
VT4025000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

131482 4-Phenylthiosemicarbazide, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?