• USA Home
  • 79226 - Tributylphenylstannane

EMAIL THIS PAGE TO A FRIEND
79226 Sigma-Aldrich

Tributylphenylstannane

purum, ≥97.0% (GC)

Synonym: Phenyltributylstannane, Phenyltributyltin, Tributylphenyltin

  • CAS Number 960-16-7

  • Linear Formula C6H5Sn[(CH2)3CH3]3

  • Molecular Weight 367.16

  •  Beilstein Registry Number 3610571

  •  MDL number MFCD00134394

  •  PubChem Substance ID 329768322

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organotin, Organotins
InChI Key   SYUVAXDZVWPKSI-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.516(lit.)
bp   125-128 °C/0.14 mmHg(lit.)
density   1.125 g/mL at 25 °C(lit.)

Description

Other Notes

Stannane used for the transfer of a phenyl group in palladium-catalyzed reactions

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2788 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?