• USA Home
  • 79233 - (S)-(+)-α-(Trifluoromethyl)benzyl alcohol

EMAIL THIS PAGE TO A FRIEND
79233 Sigma-Aldrich

(S)-(+)-α-(Trifluoromethyl)benzyl alcohol

puriss., ≥99.0% (sum of enantiomers, GC)

Synonym: (+)-Phenyl(trifluoromethyl)carbinol, (S)-(+)-1-Phenyl-2,2,2-trifluoroethanol

  • CAS Number 340-06-7

  • Linear Formula C6H5CH(CF3)OH

  • Molecular Weight 176.14

  •  Beilstein Registry Number 2327547

  •  EC Number 206-430-7

  •  MDL number MFCD00077845

Purchase

Properties

Related Categories Alcohols, Analytical Reagents, Analytical/Chromatography, Asymmetric Synthesis, Chemical Synthesis,
InChI Key   VNOMEAQPOMDWSR-ZETCQYMHSA-N
grade   puriss.
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]20/D 31.5±1.5°, neat
optical purity   enantiomeric ratio: ≥98:2 (GC)
refractive index   n20/D 1.462(lit.)
  n20/D 1.462
bp   73-76 °C/9 mmHg(lit.)
density   1.3 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
183.2 °F
Flash Point(C) 
84 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?