• USA Home
  • 79305 - Phleomycin from Streptomyces verticillus

EMAIL THIS PAGE TO A FRIEND
79305 Sigma-Aldrich

Phleomycin from Streptomyces verticillus

Purchase

Properties

Related Categories Antibacterial, Antibiotics, Antibiotics A to Z, Antibiotics N-S, Antibiotics by Application,
impurities   ~1% Cu
solubility   H2O: 10 mg/mL clear, blue
Mode of action   DNA synthesis | interferes
antibiotic activity spectrum   fungi
storage temp.   2-8°C

Description

Other Notes

Cleavage of cellular and extracellular Saccharomyces cerevisiae DNA

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
SY0525000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P9564 Phleomycin from Streptomyces verticillus, powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?