EMAIL THIS PAGE TO A FRIEND
79715 Sigma-Aldrich

O-Phospho-L-serine

~95% (TLC)

Synonym: L-2-Amino-3-hydroxypropanoic acid 3-phosphate, L-SOP, L-Serine O-phosphate

  • CAS Number 407-41-0

  • Linear Formula (HO)2P(O)OCH2CH(NH2)CO2H

  • Molecular Weight 185.07

  •  Beilstein Registry Number 1726826

  •  EC Number 206-986-0

  •  MDL number MFCD00065935

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, I - Z, Modified Amino Acids
InChI Key   BZQFBWGGLXLEPQ-REOHCLBHSA-N
assay   ~95% (TLC)
impurities   ≤2% water
mp   190 °C(lit.)
solubility   H2O: soluble50 mg/mL, clear, colorless to slightly yellow (hot)

Description

Biochem/physiol Actions

Group III metabotropic glutamate receptor agonist (mGluR4, mGluR7, and mGluR8).

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?