EMAIL THIS PAGE TO A FRIEND
79780 Sigma-Aldrich

Phthalimide

puriss., ≥99.0% (T)

Synonym: 1,3-Dihydro-1,3-dioxoisoindole

  • CAS Number 85-41-6

  • Empirical Formula (Hill Notation) C8H5NO2

  • Molecular Weight 147.13

  •  Beilstein Registry Number 118522

  •  EC Number 201-603-3

  •  MDL number MFCD00005881

  •  PubChem Substance ID 329768623

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Chemical Synthesis, Cyclic Imides, Organic Building Blocks More...
InChI Key   XKJCHHZQLQNZHY-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (T)
impurities   ≤0.05% ash
mp   232-235 °C(lit.)
  232-236 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
TI3920000
Flash Point(F) 
302 °F
Flash Point(C) 
150 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?