EMAIL THIS PAGE TO A FRIEND
79850 Sigma-Aldrich

N-Phthaloylglycine

puriss., ≥99.0% (T)

Synonym: 1,3-Dioxo-2-isoindolineacetic acid, Phthalimidoacetic acid

  • CAS Number 4702-13-0

  • Empirical Formula (Hill Notation) C10H7NO4

  • Molecular Weight 205.17

  •  Beilstein Registry Number 184174

  •  EC Number 225-177-3

  •  MDL number MFCD00005900

  •  PubChem Substance ID 24887506

  •  eCl@ss 32160406

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Glycine, Peptide Chemistry More...
InChI Key   WQINSVOOIJDOLJ-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (T)
ign. residue   ≤0.05%
mp   193-196 °C(lit.)
  193-196 °C

Description

Other Notes

Protected glycine. The acid chloride is employed in (2+2) ketene-imine-cycloadditions

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NR3439000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?