EMAIL THIS PAGE TO A FRIEND
80191 Sigma-Aldrich

Phytol

technical, mixture of isomers (~2/1: trans/cis), ≥90% (sum of enantiomers, GC)

Synonym: 3,7,11,15-Tetramethyl-2-hexadecen-1-ol

  • CAS Number 7541-49-3

  • Linear Formula CH3[CH(CH3)(CH2)3]3C(CH3)=CHCH2OH

  • Molecular Weight 296.53

  •  Beilstein Registry Number 1726098

  •  EC Number Z-7541493

  •  MDL number MFCD00002919

  •  PubChem Substance ID 329768728

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Cell Biology, Chemical Synthesis,
InChI Key   BOTWFXYSPFMFNR-HMMYKYKNSA-N
grade   technical
assay   ≥90% (sum of enantiomers, GC)
refractive index   n20/D 1.463(lit.)
  n20/D 1.463
bp   202-204 °C/10 mmHg(lit.)
density   0.85 g/mL at 25 °C(lit.)

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

GC Analysis of Fragrance Compounds on SUPELCOWAX® 10

From our library of Articles, Sigma-Aldrich presents GC Analysis of Fragrance Compounds on SUPELCOWAX® 10
Keywords: Chromatography, Cosmetics, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?