EMAIL THIS PAGE TO A FRIEND
80240 Sigma-Aldrich

4-Picoline

technical, ≥98.0% (GC)

Synonym: 4-Methylpyridine

  • CAS Number 108-89-4

  • Empirical Formula (Hill Notation) C6H7N

  • Molecular Weight 93.13

  •  Beilstein Registry Number 104586

  •  EC Number 203-626-4

  •  MDL number MFCD00006440

Purchase

Properties

Related Categories Building Blocks, C6, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
vapor density   3.2 (vs air)
vapor pressure   4 mmHg ( 20 °C)
InChI Key   FKNQCJSGGFJEIZ-UHFFFAOYSA-N
grade   technical
assay   ≥98.0% (GC)
autoignition temp.   1000 °F
refractive index   n20/D 1.504(lit.)
  n20/D 1.506
bp   145 °C(lit.)
mp   2.4 °C(lit.)
density   0.957 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 2313 3 / PGIII
WGK Germany 
1
RTECS 
UT5425000
Flash Point(F) 
102.2 °F
Flash Point(C) 
39 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

131490 4-Methylpyridine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?