EMAIL THIS PAGE TO A FRIEND
80300 Sigma-Aldrich

Ethyl 2-picolinate

purum, ≥99.0% (GC)

Synonym: Ethyl pyridine-2-carboxylate

  • CAS Number 2524-52-9

  • Empirical Formula (Hill Notation) C8H9NO2

  • Molecular Weight 151.16

  •  Beilstein Registry Number 122485

  •  EC Number 219-758-0

  •  MDL number MFCD00006292

  •  PubChem Substance ID 329768771

Purchase

Properties

Related Categories Building Blocks, C8, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   FQYYIPZPELSLDK-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.511(lit.)
  n20/D 1.511
bp   240-241 °C(lit.)
mp   2 °C(lit.)
density   1.119 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
224.6 °F
Flash Point(C) 
107 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

E45414 Ethyl 2-picolinate, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?