EMAIL THIS PAGE TO A FRIEND
80311 Sigma-Aldrich

C1-C6 n-Paraffins

100 ppm each component in nitrogen, analytical standard

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Fuels and Hydrocarbons, Petrochemical Standards
grade   analytical standard
form   gas
shelf life   limited shelf life, expiry date on the label
concentration   100 ppm each component in nitrogen
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   multi-component solution

Description

Components

methane, ethane, propane, butane, pentane, hexane

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability


Nylon Membrane and Net Filters
Safety & Documentation

Safety Information

Symbol 
GHS04  GHS04
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1950A 2.2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?