• USA Home
  • 80373 - Fluoride, certified anion standard solution

EMAIL THIS PAGE TO A FRIEND
80373 Sigma-Aldrich

Fluoride, certified anion standard solution

1000 mg/kg F-, for ion chromatography, certified by BAM, according to ISO 17025

Synonym: Sodium fluoride solution

Purchase

Properties

grade   for ion chromatography
InChI Key   PUZPDOWCWNUUKD-UHFFFAOYSA-M
quality   certified by BAM, according to ISO 17025
concentration   1000 mg/kg F-
application(s)   ion chromatography: suitable
density   1.005 g/mL at 25 °C(lit.)
format   single component solution

Description

Analysis Note

Exact concentration, expiry date, traceability and detailed information on certification can be found on the certificate and certification report, included in each package. Content and expiry date on label.

Preparation Note

prepared with high purity NaF and high purity water (18 MOhm, filtered 0.22 μm)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

77365 Fluoride Standard for IC, TraceCERT®, 1000 mg/L fluoride in water

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?