EMAIL THIS PAGE TO A FRIEND
80456 Sigma-Aldrich

Picric acid solution

~1.3% in H2O, solution (saturated aqueous)

Synonym: 2,4,6-Trinitrophenol

  • CAS Number 88-89-1

  • Linear Formula (O2N)3C6H2OH

  • Molecular Weight 229.10

  •  Beilstein Registry Number 423400

  •  MDL number MFCD00007102

Purchase

Properties

InChI Key   OXNIZHLAWKMVMX-UHFFFAOYSA-N
form   solution (saturated aqueous)
concentration   ~1.3% in H2O
density   1.00 g/mL at 20 °C

Description

Caution

saturated solution, precipitate or colloid formation possible

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Supplemental Hazard Statements 
Explosive when dry.
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

P6744 Picric acid solution, 1.3% in H2O (saturated)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?