EMAIL THIS PAGE TO A FRIEND
80585 Sigma-Aldrich

Pinane

purum, mixture of isomers, ≥98.0% (GC)

Synonym: 2,6,6-Trimethylbicyclo[3.1.1]heptane

  • CAS Number 473-55-2

  • Empirical Formula (Hill Notation) C10H18

  • Molecular Weight 138.25

  •  Beilstein Registry Number 5237941

  •  EC Number 207-467-1

  •  MDL number MFCD00078047

  •  PubChem Substance ID 329768965

Purchase

Properties

Related Categories Alkanes, Building Blocks, Chemical Synthesis, Cyclic, Organic Building Blocks More...
InChI Key   XOKSLPVRUOBDEW-CFCGPWAMSA-N
grade   purum
assay   ≥98.0% (GC)
density   0.857 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 2319 3 / PGIII
WGK Germany 
3
Flash Point(F) 
118.4 °F
Flash Point(C) 
48 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?