EMAIL THIS PAGE TO A FRIEND
80593 Sigma-Aldrich

(1R)-(+)-cis-Pinane

analytical standard

Synonym: (1R,2S)-2,6,6-Trimethylbicyclo[3.1.1]heptane

  • CAS Number 4795-86-2

  • Empirical Formula (Hill Notation) C10H18

  • Molecular Weight 138.25

  •  Beilstein Registry Number 2036204

  •  EC Number 225-352-4

  •  MDL number MFCD00066420

  •  PubChem Substance ID 329768974

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, Chromatography,
grade   analytical standard
InChI Key   XOKSLPVRUOBDEW-DJLDLDEBSA-N
assay   ≥98.5% (sum of enantiomers, GC)
form   neat
optical activity   [α]20/D +24±1°, neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.463
bp   168-169 °C(lit.)
density   0.856 g/mL at 20 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 2319 3 / PGIII
WGK Germany 
3
Flash Point(F) 
118.4 °F
Flash Point(C) 
48 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?