EMAIL THIS PAGE TO A FRIEND
80615 Sigma-Aldrich

L-Pipecolic acid

puriss., ≥99.0% (NT)

Synonym: (S)-(−)-2-Piperidinecarboxylic acid, L-Homoproline

  • CAS Number 3105-95-1

  • Empirical Formula (Hill Notation) C6H11NO2

  • Molecular Weight 129.16

  •  Beilstein Registry Number 81093

  •  EC Number 221-462-1

  •  MDL number MFCD00005981

  •  PubChem Substance ID 329768997

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Biology, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Peptide Chemistry,
InChI Key   HXEACLLIILLPRG-YFKPBYRVSA-N
grade   puriss.
assay   ≥99.0% (NT)
optical activity   [α]20/D −26±1°, c = 4% in H2O
mp   272 °C(lit.)

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
Protocols & Articles

Articles

Proline Derivatives and Analogs

Proline is a non-polar proteinogenic amino acid that forms a tertiary amide when incorporated into peptides. It does not have a hydrogen on the amide group and therefore cannot act as a hydrogen bond...
Chemfiles Volume 5 Article 12
Keywords: Adhesion, Biochemistry, Catalysis, Deposition, Hydroxylations, Isomerizations, Nucleic acid hybridization, Pharmaceutical, Reductions, Substitutions, Type

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P2519 L-Pipecolic acid, 99% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?