EMAIL THIS PAGE TO A FRIEND
80641 Sigma-Aldrich

Piperidine

puriss. plus, ≥99.5% (GC)

Synonym: Hexahydropyridine

  • CAS Number 110-89-4

  • Empirical Formula (Hill Notation) C5H11N

  • Molecular Weight 85.15

  •  Beilstein Registry Number 102438

  •  EC Number 203-813-0

  •  MDL number MFCD00005979

  •  PubChem Substance ID 329769005

Purchase

Properties

Related Categories Building Blocks, C5 to C7, Cell Biology, Chemical Synthesis, Heterocyclic Building Blocks,
vapor density   3 (vs air)
vapor pressure   23 mmHg ( 20 °C)
InChI Key   NQRYJNQNLNOLGT-UHFFFAOYSA-N
grade   puriss. plus
assay   ≥99.5% (GC)
impurities   ≤0.5% water
refractive index   n20/D 1.452(lit.)
  n20/D 1.452
bp   106 °C(lit.)
mp   −13 °C(lit.)
solubility   organic solvents: soluble(lit.)
  water: miscible(lit.)
density   0.862 g/mL at 20 °C(lit.)

Description

Other Notes

Sales restrictions may apply

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2401 3(8) / PGI
WGK Germany 
1
RTECS 
TM3500000
Flash Point(F) 
60.8 °F
Flash Point(C) 
16 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?