• USA Home
  • 80670 - Piperazine dihydrochloride hydrate

EMAIL THIS PAGE TO A FRIEND
80670 Sigma-Aldrich

Piperazine dihydrochloride hydrate

purum, ≥97.0% (AT)

  • CAS Number 207605-49-0

  • Empirical Formula (Hill Notation) C4H10N2 · 2HCl · xH2O

  • Molecular Weight 159.06 (anhydrous basis)

  •  Beilstein Registry Number 3551215

  •  MDL number MFCD00150089

  •  PubChem Substance ID 329769024

Purchase

Properties

Related Categories Building Blocks, C4 to C8, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
InChI Key   ZQVBNUIFDLXAFI-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (AT)
impurities   ≤12% water

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?