• USA Home
  • 80805 - 1,4-Di(maleimido)butane

EMAIL THIS PAGE TO A FRIEND
80805 Sigma-Aldrich

1,4-Di(maleimido)butane

≥99.0% (HPLC)

Synonym: 1,4-Bis(maleimido)butane, BMB, N,N′-(Tetramethylene)dimaleimide

  • CAS Number 28537-70-4

  • Empirical Formula (Hill Notation) C12H12N2O4

  • Molecular Weight 248.23

  •  Beilstein Registry Number 1538100

  •  MDL number MFCD01318342

  •  PubChem Substance ID 329769053

Purchase

Properties

InChI Key   WXXSHAKLDCERGU-UHFFFAOYSA-N
assay   ≥99.0% (HPLC)

Description

Application

Linker for preparing biomimetics: J. Med. Chem. 35, 1563 (1992)

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?