• USA Home
  • 80847 - Polydisperse Particle Standard

EMAIL THIS PAGE TO A FRIEND
80847 Sigma-Aldrich

Polydisperse Particle Standard

certified reference material, Particle Size Standard, PS213, 10 x 0.25g, 10-100 μm

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Particle Size Standards, Physical Properties Standards
grade   certified reference material
form   particles
mfr. no.   Whitehouse Scientific Ltd PS213
particle size   10-100 μm
format   neat

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?