EMAIL THIS PAGE TO A FRIEND
80850 Sigma-Aldrich

Piperonylbutoxide

technical, ≥90% (GC)

Synonym: 2-(2-Butoxyethoxy)ethyl (6-propylpiperonyl) ether, 4,5-Methylenedioxy-2-propylbenzyldiethyleneglycol butyl ether

  • CAS Number 51-03-6

  • Empirical Formula (Hill Notation) C19H30O5

  • Molecular Weight 338.44

  •  Beilstein Registry Number 288063

  •  EC Number 200-076-7

  •  MDL number MFCD00005842

  •  PubChem Substance ID 24887361

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Protected Alcohols/Phenols More...
InChI Key   FIPWRIJSWJWJAI-UHFFFAOYSA-N
grade   technical
assay   ≥90% (GC)
refractive index   n20/D 1.498(lit.)
  n20/D 1.500
bp   155 °C/0.3 mmHg(lit.)
density   1.059 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
XS8050000
Flash Point(F) 
339.8 °F
Flash Point(C) 
171 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

291102 Piperonylbutoxide, technical grade, 90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?